Assay Details
| Gene Symbol: SFR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203747.1 | NP_001190676.1 | 11 - 12 | 2705 | 67 | |
| NM_001340519.1 | NP_001329057.1 | 11 - 12 | 2751 | 67 | ||
| NM_116730.5 | NP_192401.5 | 11 - 12 | 2418 | 67 | ||
More Information
Set Membership: |
|

SFR6
Genomic Map