Assay Details
| Gene Symbol: DES-1-LIKE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116731.4 | NP_192402.1 | 1 - 2 | 363 | 77 | |
| GenBank mRNA | AF220201.1 | - | 1 - 2 | 283 | 77 | |
| AK221700.1 | - | 1 - 2 | 261 | 77 | ||
More Information
Set Membership: |
|

DES-1-LIKE
Genomic Map