Assay Details
| Gene Symbol: LpxD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340544.1 | NP_001328366.1 | 7 - 8 | 929 | 65 | |
| NM_001340545.1 | NP_001328365.1 | 8 - 9 | 729 | 65 | ||
| NM_116760.2 | NP_192430.2 | 8 - 9 | 711 | 65 | ||
More Information
Set Membership: |
|

LpxD1
Genomic Map