Assay Details
| Gene Symbol: AT4G08310 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116900.5 | NP_192571.1 | 7 - 8 | 1390 | 85 | |
| GenBank mRNA | AY091128.1 | - | 7 - 8 | 1211 | 85 | |
| AY142689.1 | - | 7 - 8 | 1202 | 85 | ||
More Information
Set Membership: |
|

AT4G08310
Genomic Map