Assay Details
| Gene Symbol: TPR8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036517.3 | NP_001031594.1 | 7 - 8 | 973 | 78 | |
| NM_116901.4 | NP_192572.2 | 7 - 8 | 1068 | 78 | ||
More Information
Set Membership: |
|

TPR8
Genomic Map