Assay Details
| Gene Symbol: SAPX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084887.2 | NP_001078356.2 | 2 - 3 | 593 | 144 | |
| NM_001203761.1 | NP_001190690.1 | 2 - 3 | 596 | 144 | ||
| NM_001340582.1 | NP_001319883.1 | 2 - 3 | 630 | 144 | ||
| NM_116908.3 | NP_192579.1 | 2 - 3 | 608 | 144 | ||
| NM_202791.3 | NP_974520.1 | 2 - 3 | 596 | 144 | ||
| GenBank mRNA | AY056319.1 | - | 2 - 3 | 534 | 144 | |
| AY114065.1 | - | 2 - 3 | 502 | 144 | ||
More Information
Set Membership: |
|

SAPX
Genomic Map