Assay Details
| Gene Symbol: SAPX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084887.2 | NP_001078356.2 | 4 - 5 | 784 | 67 | |
| NM_001203761.1 | NP_001190690.1 | 4 - 5 | 787 | 67 | ||
| NM_001340582.1 | NP_001319883.1 | 4 - 5 | 821 | 67 | ||
| NM_116908.3 | NP_192579.1 | 4 - 5 | 799 | 67 | ||
| NM_202791.3 | NP_974520.1 | 4 - 5 | 787 | 67 | ||
| GenBank mRNA | AY056319.1 | - | 4 - 5 | 725 | 67 | |
| AY114065.1 | - | 4 - 5 | 693 | 67 | ||
More Information
Set Membership: |
|

SAPX
Genomic Map