Assay Details
| Gene Symbol: SAPX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084887.2 | NP_001078356.2 | 9 - 10 | 1195 | 148 | |
| NM_001203761.1 | NP_001190690.1 | 8 - 9 | 1120 | 148 | ||
| NM_001340582.1 | NP_001319883.1 | 9 - 10 | 1232 | 148 | ||
| NM_116908.3 | NP_192579.1 | 9 - 10 | 1210 | 148 | ||
| NM_202791.3 | NP_974520.1 | 9 - 10 | 1198 | 151 | ||
| GenBank mRNA | AY056319.1 | - | 9 - 10 | 1136 | 148 | |
More Information
Set Membership: |
|

SAPX
Genomic Map