Assay Details
| Gene Symbol: AT4G08540 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116923.4 | NP_192594.2 | 2 - 3 | 306 | 130 | |
| GenBank mRNA | AY150481.1 | - | 2 - 3 | 167 | 130 | |
More Information
Set Membership: |
|

AT4G08540
Genomic Map