Assay Details
| Gene Symbol: TL29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340606.1 | NP_001329837.1 | 4 - 5 | 713 | 129 | |
| NM_001340607.1 | NP_001329836.1 | 5 - 6 | 660 | 129 | ||
| NM_116970.3 | NP_192640.1 | 5 - 6 | 550 | 129 | ||
| GenBank mRNA | AF370534.1 | - | 5 - 6 | 480 | 129 | |
| AY072503.1 | - | 5 - 6 | 426 | 129 | ||
More Information
Set Membership: |
|

TL29
Genomic Map