Assay Details
| Gene Symbol: TL29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340606.1 | NP_001329837.1 | 8 - 9 | 1229 | 88 | |
| NM_001340607.1 | NP_001329836.1 | 9 - 10 | 1176 | 88 | ||
| NM_116970.3 | NP_192640.1 | 9 - 10 | 1066 | 88 | ||
| GenBank mRNA | AF370534.1 | - | 9 - 10 | 996 | 88 | |
| AY072503.1 | - | 9 - 10 | 942 | 88 | ||
More Information
Set Membership: |
|

TL29
Genomic Map