Assay Details
| Gene Symbol: AT4G09070 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116976.1 | NP_192646.1 | 1 - 2 | 426 | 122 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT4G09070| Gene Symbol: AT4G09070 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116976.1 | NP_192646.1 | 1 - 2 | 426 | 122 | |
Set Membership: |
|