Assay Details
| Gene Symbol: SULTR3;5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121965.4 | NP_568377.1 | 1 - 2 | 481 | 66 | |
| GenBank mRNA | AB061739.1 | - | 1 - 2 | 431 | 66 | |
| AK226785.1 | - | 1 - 2 | 439 | 66 | ||
More Information
Set Membership: |
|

SULTR3;5
Genomic Map