Assay Details
| Gene Symbol: S1P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343620.1 | NP_001331147.1 | 2 - 3 | 808 | 70 | |
| NM_121971.3 | NP_197467.1 | 2 - 3 | 804 | 70 | ||
| GenBank mRNA | AK227193.1 | - | 2 - 3 | 788 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001343620.1 transcripts.
Set Membership: |
|

S1P
Genomic Map