Assay Details
| Gene Symbol: S1P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343619.1 | NP_001331146.1 | 5 - 6 | 1313 | 62 | |
| NM_001343620.1 | NP_001331147.1 | 5 - 6 | 1280 | 62 | ||
| NM_121971.3 | NP_197467.1 | 5 - 6 | 1266 | 62 | ||
| GenBank mRNA | AK227193.1 | - | 5 - 6 | 1250 | 62 | |
More Information
Set Membership: |
|

S1P
Genomic Map