Assay Details
| Gene Symbol: S1P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343619.1 | NP_001331146.1 | 9 - 10 | 3198 | 74 | |
| NM_001343620.1 | NP_001331147.1 | 9 - 10 | 3165 | 74 | ||
| NM_121971.3 | NP_197467.1 | 9 - 10 | 3151 | 74 | ||
| GenBank mRNA | AK227193.1 | - | 9 - 10 | 3135 | 74 | |
More Information
Set Membership: |
|

S1P
Genomic Map