Assay Details
| Gene Symbol: AT5G19730 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121978.4 | NP_197474.1 | 4 - 5 | 1290 | 78 | |
| GenBank mRNA | AY065431.1 | - | 4 - 5 | 1107 | 78 | |
| AY096559.1 | - | 4 - 5 | 1003 | 78 | ||
More Information
Set Membership: |
|

AT5G19730
Genomic Map