Assay Details
| Gene Symbol: AT5G19760 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121981.5 | NP_197477.1 | 3 - 4 | 472 | 79 | |
| GenBank mRNA | AF360153.1 | - | 3 - 4 | 372 | 79 | |
| AJ311780.1 | - | 3 - 4 | 356 | 79 | ||
| AK226470.1 | - | 3 - 4 | 377 | 79 | ||
| AY056307.1 | - | 3 - 4 | 263 | 79 | ||
| AY085901.1 | - | 3 - 4 | 372 | 79 | ||
More Information
Set Membership: |
|

AT5G19760
Genomic Map