Assay Details
| Gene Symbol: AT5G19880 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121995.2 | NP_197488.1 | 1 - 2 | 277 | 108 | |
| GenBank mRNA | DQ446972.1 | - | 1 - 2 | 211 | 108 | |
More Information
Set Membership: |
|

AT5G19880
Genomic Map