Assay Details
| Gene Symbol: MED31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203413.1 | NP_001190342.1 | 1 - 2 | 218 | 67 | |
| NM_121998.5 | NP_197491.2 | 1 - 2 | 136 | 67 | ||
| GenBank mRNA | AY072222.1 | - | 1 - 2 | 88 | 67 | |
| BT002744.1 | - | 1 - 2 | 49 | 67 | ||
More Information
Set Membership: |
|

MED31
Genomic Map