Assay Details
| Gene Symbol: PDIL1-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104105.3 | NP_175636.2 | 11 - 12 | 1729 | 66 | |
| GenBank mRNA | BT030322.1 | - | 11 - 12 | 1517 | 66 | |
More Information
Set Membership: |
|

PDIL1-5
Genomic Map