Assay Details
| Gene Symbol: BGLU18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036098.2 | NP_001031175.1 | 5 - 6 | 544 | 126 | |
| NM_001198275.1 | NP_001185204.1 | 5 - 6 | 552 | 126 | ||
| NM_104118.3 | NP_175649.1 | 5 - 6 | 706 | 126 | ||
| GenBank mRNA | AY039855.1 | - | 5 - 6 | 521 | 126 | |
| AY056415.1 | - | 5 - 6 | 521 | 126 | ||
| BT000515.1 | - | 5 - 6 | 445 | 126 | ||
More Information
Set Membership: |
|

BGLU18
Genomic Map