Assay Details
| Gene Symbol: BGLU18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036098.2 | NP_001031175.1 | 7 - 8 | 1021 | 103 | |
| NM_001198275.1 | NP_001185204.1 | 7 - 8 | 1029 | 103 | ||
| NM_104118.3 | NP_175649.1 | 7 - 8 | 1183 | 103 | ||
| GenBank mRNA | AY039855.1 | - | 7 - 8 | 998 | 103 | |
| AY056415.1 | - | 7 - 8 | 998 | 103 | ||
| BT000515.1 | - | 7 - 8 | 922 | 103 | ||
More Information
Set Membership: |
|

BGLU18
Genomic Map