Assay Details
| Gene Symbol: AT1G52590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104137.2 | NP_564611.1 | 2 - 3 | 287 | 101 | |
| GenBank mRNA | AK117491.1 | - | 2 - 3 | 252 | 101 | |
| BT005075.1 | - | 2 - 3 | 237 | 101 | ||
More Information
Set Membership: |
|

AT1G52590
Genomic Map