Assay Details
| Gene Symbol: AT1G52590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104137.2 | NP_564611.1 | 4 - 5 | 481 | 95 | |
| GenBank mRNA | AK117491.1 | - | 4 - 5 | 446 | 95 | |
| BT005075.1 | - | 4 - 5 | 431 | 95 | ||
More Information
Set Membership: |
|

AT1G52590
Genomic Map