Assay Details
| Gene Symbol: AT1G52810 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104159.1 | NP_175690.1 | 2 - 3 | 506 | 111 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G52810| Gene Symbol: AT1G52810 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104159.1 | NP_175690.1 | 2 - 3 | 506 | 111 | |
Set Membership: |
|