Assay Details
| Gene Symbol: SPA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333576.1 | NP_001321533.1 | 2 - 3 | 974 | 149 | |
| NM_001333578.1 | NP_001321534.1 | 1 - 2 | 1024 | 149 | ||
| NM_104188.4 | NP_175717.1 | 2 - 3 | 1078 | 149 | ||
| NM_179471.2 | NP_849802.1 | 1 - 2 | 1052 | 149 | ||
| GenBank mRNA | AY040023.1 | - | 2 - 3 | 721 | 149 | |
| AY133883.1 | - | 1 - 2 | 628 | 149 | ||
More Information
Set Membership: |
|

SPA4
Genomic Map