Assay Details
| Gene Symbol: SPA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333576.1 | NP_001321533.1 | 6 - 7 | 2474 | 84 | |
| NM_001333577.1 | NP_001321532.1 | 6 - 7 | 2318 | 84 | ||
| NM_104188.4 | NP_175717.1 | 6 - 7 | 2578 | 84 | ||
| NM_179471.2 | NP_849802.1 | 5 - 6 | 2552 | 84 | ||
| GenBank mRNA | AY040023.1 | - | 6 - 7 | 2221 | 84 | |
| AY133883.1 | - | 5 - 6 | 2128 | 84 | ||
More Information
Set Membership: |
|

SPA4
Genomic Map