Assay Details
| Gene Symbol: GRI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104192.3 | NP_175721.1 | 1 | 489 | 97 | |
| GenBank mRNA | AK176040.1 | - | 1 | 489 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GRI| Gene Symbol: GRI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104192.3 | NP_175721.1 | 1 | 489 | 97 | |
| GenBank mRNA | AK176040.1 | - | 1 | 489 | 97 | |
Set Membership: |
|