Assay Details
| Gene Symbol: MUM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104228.3 | NP_564633.2 | 2 - 3 | 1867 | 112 | |
| GenBank mRNA | AJ565874.1 | - | 1 - 2 | 1558 | 112 | |
More Information
Set Membership: |
|

MUM4
Genomic Map