Assay Details
| Gene Symbol: CID8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036105.1 | NP_001031182.1 | 7 - 8 | 937 | 77 | |
| NM_104243.4 | NP_175769.1 | 7 - 8 | 1127 | 77 | ||
| GenBank mRNA | BT014885.1 | - | 7 - 8 | 751 | 77 | |
More Information
Set Membership: |
|

CID8
Genomic Map