Assay Details
| Gene Symbol: CID8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036105.1 | NP_001031182.1 | 9 - 10 | 1072 | 66 | |
| NM_104243.4 | NP_175769.1 | 9 - 10 | 1262 | 66 | ||
| GenBank mRNA | BT014885.1 | - | 9 - 10 | 886 | 66 | |
More Information
Set Membership: |
|

CID8
Genomic Map