Assay Details
| Gene Symbol: MSRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001124012.1 | NP_001117484.1 | 1 - 2 | 234 | 95 | |
| NM_104245.4 | NP_564640.2 | 1 - 2 | 244 | 95 | ||
| GenBank mRNA | BT030636.1 | - | 1 - 2 | 188 | 95 | |
More Information
Set Membership: |
|

MSRB1
Genomic Map