Assay Details
| Gene Symbol: SRF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084247.2 | NP_001077716.1 | 8 - 9 | 1019 | 60 | |
| NM_104251.4 | NP_175777.1 | 8 - 9 | 942 | 60 | ||
| GenBank mRNA | AK227057.1 | - | 8 - 9 | 695 | 60 | |
| AY518291.1 | - | 8 - 9 | 723 | 60 | ||
| BT010600.1 | - | 8 - 9 | 653 | 60 | ||
| FJ708659.1 | - | 8 - 9 | 653 | 60 | ||
More Information
Set Membership: |
|

SRF6
Genomic Map