Assay Details
| Gene Symbol: PME1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104261.3 | NP_175787.1 | 1 - 2 | 1314 | 88 | |
| GenBank mRNA | AK226427.1 | - | 1 - 2 | 1139 | 88 | |
| AY054197.1 | - | 1 - 2 | 1126 | 88 | ||
| X81585.1 | - | 1137 | 88 | |||
More Information
Set Membership: |
|

PME1
Genomic Map