Assay Details
| Gene Symbol: RSH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104291.8 | NP_564652.2 | 1 - 2 | 1094 | 127 | |
| GenBank mRNA | AK227169.1 | - | 1 - 2 | 999 | 127 | |
More Information
Set Membership: |
|

RSH3
Genomic Map