Assay Details
| Gene Symbol: CID3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104295.4 | NP_175819.1 | 4 - 5 | 695 | 66 | |
| GenBank mRNA | AY136398.1 | - | 4 - 5 | 586 | 66 | |
| BT000204.1 | - | 3 - 4 | 378 | 66 | ||
More Information
Set Membership: |
|

CID3
Genomic Map