Assay Details
| Gene Symbol: CID3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104295.4 | NP_175819.1 | 7 - 8 | 1290 | 86 | |
| GenBank mRNA | AY136398.1 | - | 7 - 8 | 1181 | 86 | |
| BT000204.1 | - | 6 - 7 | 973 | 86 | ||
More Information
Set Membership: |
|

CID3
Genomic Map