Assay Details
| Gene Symbol: AT1G54200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104298.4 | NP_175822.1 | 1 | 1325 | 103 | |
| GenBank mRNA | BT029487.1 | - | 1 | 935 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G54200| Gene Symbol: AT1G54200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104298.4 | NP_175822.1 | 1 | 1325 | 103 | |
| GenBank mRNA | BT029487.1 | - | 1 | 935 | 103 | |
Set Membership: |
|