Assay Details
| Gene Symbol: EIF4A-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198291.2 | NP_001185220.1 | 2 - 3 | 288 | 102 | |
| NM_104305.4 | NP_175829.1 | 2 - 3 | 487 | 102 | ||
| GenBank mRNA | AF386923.1 | - | 2 - 3 | 215 | 102 | |
| AF428462.1 | - | 2 - 3 | 212 | 102 | ||
| AK226344.1 | - | 2 - 3 | 221 | 102 | ||
| AY087965.1 | - | 2 - 3 | 214 | 102 | ||
| BT008504.1 | - | 1 - 2 | 70 | 102 | ||
| X65053.1 | - | 220 | 102 | |||
More Information
Set Membership: |
|

EIF4A-2
Genomic Map