Assay Details
| Gene Symbol: TAF6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036110.1 | NP_001031187.2 | 3 - 4 | 304 | 98 | |
| NM_001198292.1 | NP_001185221.1 | 3 - 4 | 218 | 98 | ||
| NM_001333642.1 | NP_001319224.1 | 2 - 3 | 370 | 98 | ||
| NM_001333643.1 | NP_001322136.1 | 2 - 3 | 414 | 98 | ||
| NM_104314.6 | NP_175838.4 | 3 - 4 | 346 | 98 | ||
| NM_202295.2 | NP_974024.2 | 2 - 3 | 399 | 98 | ||
More Information
Set Membership: |
|

TAF6B
Genomic Map