Assay Details
| Gene Symbol: TAF6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036110.1 | NP_001031187.2 | 4 - 5 | 406 | 86 | |
| NM_001198292.1 | NP_001185221.1 | 4 - 5 | 320 | 86 | ||
| NM_001333642.1 | NP_001319224.1 | 3 - 4 | 472 | 86 | ||
| NM_001333643.1 | NP_001322136.1 | 3 - 4 | 516 | 86 | ||
| NM_104314.6 | NP_175838.4 | 4 - 5 | 448 | 86 | ||
| NM_202295.2 | NP_974024.2 | 3 - 4 | 501 | 86 | ||
More Information
Set Membership: |
|

TAF6B
Genomic Map