Assay Details
| Gene Symbol: TAF6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036110.1 | NP_001031187.2 | 9 - 10 | 957 | 89 | |
| NM_001198292.1 | NP_001185221.1 | 9 - 10 | 892 | 89 | ||
| NM_001333642.1 | NP_001319224.1 | 8 - 9 | 1044 | 89 | ||
| NM_001333643.1 | NP_001322136.1 | 8 - 9 | 1088 | 89 | ||
| NM_104314.6 | NP_175838.4 | 9 - 10 | 1020 | 89 | ||
| NM_202295.2 | NP_974024.2 | 8 - 9 | 1073 | 89 | ||
More Information
Set Membership: |
|

TAF6B
Genomic Map