Assay Details
| Gene Symbol: ING2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104317.3 | NP_564658.2 | 2 - 3 | 195 | 112 | |
| NM_202296.2 | NP_974025.1 | 2 - 3 | 195 | 112 | ||
| NM_202297.3 | NP_974026.1 | 2 - 3 | 281 | 112 | ||
| NM_202298.3 | NP_974027.1 | 2 - 3 | 281 | 112 | ||
| GenBank mRNA | AK316931.1 | - | 2 - 3 | 188 | 112 | |
| AY099578.1 | - | 2 - 3 | 143 | 112 | ||
| BT002159.1 | - | 2 - 3 | 114 | 112 | ||
More Information
Set Membership: |
|

ING2
Genomic Map