Assay Details
| Gene Symbol: AT1G54450 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104323.4 | NP_175847.1 | 1 - 2 | 367 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G54450| Gene Symbol: AT1G54450 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104323.4 | NP_175847.1 | 1 - 2 | 367 | 65 | |
Set Membership: |
|