Assay Details
| Gene Symbol: AT1G54460 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104324.4 | NP_564659.1 | 4 - 5 | 705 | 68 | |
| GenBank mRNA | AF361626.1 | - | 4 - 5 | 680 | 68 | |
| AY055093.1 | - | 3 - 4 | 540 | 68 | ||
More Information
Set Membership: |
|

AT1G54460
Genomic Map