Assay Details
| Gene Symbol: XRN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333652.1 | NP_001321327.1 | 14 - 15 | 2000 | 99 | |
| NM_104327.4 | NP_175851.1 | 14 - 15 | 1795 | 99 | ||
| GenBank mRNA | AF286718.1 | - | 14 - 15 | 1714 | 99 | |
| AY064012.1 | - | 14 - 15 | 1711 | 99 | ||
| AY091411.1 | - | 14 - 15 | 1604 | 99 | ||
| BT026022.1 | - | 14 - 15 | 1690 | 99 | ||
More Information
Set Membership: |
|

XRN4
Genomic Map