Assay Details
| Gene Symbol: XRN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104327.4 | NP_175851.1 | 1 - 2 | 398 | 119 | |
| GenBank mRNA | AF286718.1 | - | 1 - 2 | 317 | 119 | |
| AY064012.1 | - | 1 - 2 | 314 | 119 | ||
| AY091411.1 | - | 1 - 2 | 207 | 119 | ||
| BT026022.1 | - | 1 - 2 | 293 | 119 | ||
More Information
Set Membership: |
|

XRN4
Genomic Map