Assay Details
| Gene Symbol: AT1G54520 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104330.4 | NP_564660.1 | 2 - 3 | 918 | 58 | |
| GenBank mRNA | AY085198.1 | - | 2 - 3 | 818 | 58 | |
| AY093070.1 | - | 2 - 3 | 816 | 58 | ||
More Information
Set Membership: |
|

AT1G54520
Genomic Map