Assay Details
| Gene Symbol: AT1G54520 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104330.4 | NP_564660.1 | 3 - 4 | 971 | 67 | |
| GenBank mRNA | AY085198.1 | - | 3 - 4 | 871 | 67 | |
| AY093070.1 | - | 3 - 4 | 869 | 67 | ||
More Information
Set Membership: |
|

AT1G54520
Genomic Map